
4-[(1,1-Dioxidothiomorpholin-4-yl)methyl]aniline 90%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:263339-24-8
:
Formula:C₁₁H₁₆N₂O₂S
MDL Number:MFCD03407300
:240.322 g/mol
Melting Pt:156 °C
Storage Temperature:Refrigerator
UN:3259
ADR:8,III
:GHS05
Frequently Bought Together