
3-(Naphth-1-yl)-1H-pyrazole
Catalog # APOSOR2574-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:194.232 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:150433-19-5
Formula:C₁₃H₁₀N₂
MDL Number:MFCD01569443