Order Entry
Czech Republic
ContactUsLinkComponent
3-Chloro-4-(phenylthio)tetrahydro-1H-1lambda~6~-thiophene-1,1-dione
Irritant
3-Chloro-4-(phenylthio)tetrahydro-1H-1lambda~6~-thiophene-1,1-dione
Catalog # APOSOR25447-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Chloro-4-(phenylthio)tetrahydro-1H-1lambda~6~-thiophene-1,1-dione
Catalog # APOSOR25447-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :262.776 g/mol
Melting Pt:78…80 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:15507-87-6
Formula:C₁₀H₁₁ClO₂S₂
MDL Number:MFCD00174266