Order Entry
Czech Republic
ContactUsLinkComponent
6,6-Dimethyl-3-(methylthio)-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carbonitrile
Irritant
6,6-Dimethyl-3-(methylthio)-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carbonitrile
Catalog # APOSOR25283-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6,6-Dimethyl-3-(methylthio)-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carbonitrile
Catalog # APOSOR25283-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:169…171 °C
Storage Temperature:Refrigerator
UN:3276
ADR:6.1,III
 :251.368 g/mol
 :GHS07
CAS Number:175202-50-3
Formula:C₁₂H₁₃NOS₂
MDL Number:MFCD00084980