2-({[2-(Ethylthio)-3-pyridyl]carbonyl}amino)-4-(methylthio)butanoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175201-66-8
:
Formula:C₁₃H₁₈N₂O₃S₂
MDL Number:MFCD03407299
:314.424 g/mol
Storage Temperature:Ambient
Frequently Bought Together