Order Entry
Czech Republic
ContactUsLinkComponent
4-Cyano-3-(4-methoxyphenyl)-5-(methylthio)thiophene-2-carboxylic acid
4-Cyano-3-(4-methoxyphenyl)-5-(methylthio)thiophene-2-carboxylic acid
Catalog # APOSOR2466-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Cyano-3-(4-methoxyphenyl)-5-(methylthio)thiophene-2-carboxylic acid
Catalog # APOSOR2466-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :305.372 g/mol
Melting Pt:209…213 °C
Storage Temperature:Ambient
CAS Number:116493-07-3
Formula:C₁₄H₁₁NO₃S₂
MDL Number:MFCD00277044