
5-Chloropyrazolo[1,5-a]thieno[2,3-e]pyrimidine-3-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:680210-93-9
:
Formula:C₉H₄ClN₃O₂S
MDL Number:MFCD00206489
:253.665 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together