
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carboxaldehyde oxime
Catalog # APOSOR24172-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:173.6 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:590410-59-6
Formula:C₆H₈ClN₃O
MDL Number:MFCD00121970