Order Entry
Czech Republic
ContactUsLinkComponent
5-Chloro-1,3-dimethyl-4-(tetrahydro-1H-pyrrol-1-ylsulphonyl)-1H-pyrazole
Irritant
5-Chloro-1,3-dimethyl-4-(tetrahydro-1H-pyrrol-1-ylsulphonyl)-1H-pyrazole
Catalog # APOSOR23903-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Chloro-1,3-dimethyl-4-(tetrahydro-1H-pyrrol-1-ylsulphonyl)-1H-pyrazole
Catalog # APOSOR23903-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :263.744 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:647825-38-5
Formula:C₉H₁₄ClN₃O₂S
MDL Number:MFCD00113247