
2-(4-Morpholino)benzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:42106-48-9
:
Formula:C₁₁H₁₃NO₃
MDL Number:MFCD00527976
Melting Pt:151…159 °C
:207.226 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together