Order Entry
Czech Republic
ContactUsLinkComponent
5-(Hydroxymethyl)-4-methyl-2-phenylpyrimidine
Irritant
5-(Hydroxymethyl)-4-methyl-2-phenylpyrimidine
Catalog # APOSOR23333-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-(Hydroxymethyl)-4-methyl-2-phenylpyrimidine
Catalog # APOSOR23333-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :200.236 g/mol
Melting Pt:84.5…86 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:342405-27-0
Formula:C₁₂H₁₂N₂O
MDL Number:MFCD02682074