
4-(1H-Pyrazol-1-yl)benzenesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:18336-39-5
:
Formula:C₉H₇ClN₂O₂S
MDL Number:MFCD02681937
Melting Pt:108…109 °C
UN:3261
ADR:8,III
:242.682 g/mol
:GHS05
Storage Temperature:Ambient
Frequently Bought Together