Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,9-Diazaspiro[5.5]undecan-2-one hydrochloride
1,9-Diazaspiro[5.5]undecan-2-one hydrochloride 95%
1,9-Diazaspiro[5.5]undecan-2-one hydrochloride 95%
1,9-Diazaspiro[5.5]undecan-2-one hydrochloride 95%
APOSOR2327-100MG
:
Apollo Scientific
:
1,9-Diazaspiro[5.5]undecan-2-one hydrochloride 95%
APOSOR2327-100MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 mg
Seq:
2440
:
:
>300 °C
:
204.697 g/mol
:
GHS07
:
Ambient
:
1171417-47-2
:
C₉H₁₆N₂O·ClH
:
MFCD09998743