
4-(2,3-Dihydro-1,4-benzodioxin-6-yl)butanoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14939-93-6
:
Formula:C₁₂H₁₄O₄
MDL Number:MFCD00276566
:222.237 g/mol
Melting Pt:76…78 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together