Order Entry
Czech Republic
ContactUsLinkComponent
(2S,4R)-4-Aminopyrrolidine-2-carboxylic acid, 4-BOC, N1-FMOC protected 97%
(2S,4R)-4-Aminopyrrolidine-2-carboxylic acid, 4-BOC, N1-FMOC protected 97%
Catalog # APOSOR2235-250MG
Supplier:  Apollo Scientific
CAS Number:  
(2S,4R)-4-Aminopyrrolidine-2-carboxylic acid, 4-BOC, N1-FMOC protected 97%
Catalog # APOSOR2235-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :452.5 g/mol
Storage Temperature:Ambient
CAS Number:273222-06-3
Formula:C₂₅H₂₈N₂O₆
MDL Number:MFCD01860707