
3-Methyl-N-[(1E)-3-oxo-3-phenylprop-1-en-1-yl]pyridinium chloride
Catalog # APOSOR22247-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Typ balení:Bottle
- Bal.:500 mg
- Sekvence:2725
Specifications
About this item
:
:259.731 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:19518-26-4
Formula:C₁₅H₁₄NO·Cl
MDL Number:MFCD00204657