2-(4-aminophenoxy)-2-methylpropanoic acid 95+%
Catalog # APOSOR21768-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:195.215 g/mol
Storage Temperature:Ambient
CAS Number:117011-70-8
Formula:C₁₀H₁₃NO₃
MDL Number:MFCD00084883