Order Entry
Czech Republic
ContactUsLinkComponent
5,6-Dimethyl-1-indanone 96%
Irritant
5,6-Dimethyl-1-indanone 96%
Catalog # APOSOR19567-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5,6-Dimethyl-1-indanone 96%
Catalog # APOSOR19567-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :160.212 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:16440-97-4
Formula:C₁₁H₁₂O
MDL Number:MFCD00666126