Order Entry
Czech Republic
ContactUsLinkComponent
2-Isobutyrylcyclohexan-1-one 97%
Irritant
2-Isobutyrylcyclohexan-1-one 97%
Catalog # APOSOR19543-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Isobutyrylcyclohexan-1-one 97%
Catalog # APOSOR19543-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :168.233 g/mol
Boiling Pt:63…64 °C
Melting Pt:38 °C
Density:1.0076 g/cm³ (25 °C)
Flash Pt:104.4 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:39207-65-3
Formula:C₁₀H₁₆O₂
MDL Number:MFCD00466185