tert-Butyl 4-(3,4-diaminophenyl)piperazine-1-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:193902-99-7
:
Formula:C₁₅H₂₄N₄O₂
MDL Number:MFCD12022584
:292.377 g/mol
Storage Temperature:Ambient
Frequently Bought Together