4-Hydroxy-6-methylquinoline-3-carboxylic acid
Catalog # APOSOR183475-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:203.194 g/mol
Storage Temperature:Ambient
CAS Number:35973-18-3
Formula:C₁₁H₉NO₃
MDL Number:MFCD00454147