N-[2-(1H-Pyrazol-3-yl)benzofuran-3-yl]benzamide
Catalog # APOSOR183468-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:303.315 g/mol
Storage Temperature:Ambient
CAS Number:1401786-32-0
Formula:C₁₈H₁₃N₃O₂
MDL Number:MFCD11054361