3-(2,3-Dihydrobenzo[b]furan-5-yl)-1H-pyrazole
Catalog # APOSOR183459-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:186.21 g/mol
Storage Temperature:Ambient
CAS Number:1005923-72-7
Formula:C₁₁H₁₀N₂O
MDL Number:MFCD00204129