Order Entry
Czech Republic
ContactUsLinkComponent
3-(2,3-Dihydro-1,4-benzodioxin-6-yl)-1H-pyrazole
3-(2,3-Dihydro-1,4-benzodioxin-6-yl)-1H-pyrazole
Catalog # APOSOR183458-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(2,3-Dihydro-1,4-benzodioxin-6-yl)-1H-pyrazole
Catalog # APOSOR183458-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :202.209 g/mol
Storage Temperature:Ambient
CAS Number:1019007-45-4
Formula:C₁₁H₁₀N₂O₂
MDL Number:MFCD03618499