3-Phenyl-5-(1H-pyrazol-3-yl)isoxazole
Catalog # APOSOR183449-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:211.219 g/mol
Storage Temperature:Ambient
CAS Number:264225-67-4
Formula:C₁₂H₉N₃O
MDL Number:MFCD00832976