4-[4-(1H-Pyrazol-3-yl)phenyl]sulphonylmorpholine
Catalog # APOSOR183440-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:293.342 g/mol
Storage Temperature:Ambient
CAS Number:1019011-88-1
Formula:C₁₃H₁₅N₃O₃S
MDL Number:MFCD07774261