N,N-Dimethyl-4-(1H-pyrazol-3-yl)benzenesulphonamide
Catalog # APOSOR183438-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:251.305 g/mol
Storage Temperature:Ambient
CAS Number:1019011-14-3
Formula:C₁₁H₁₃N₃O₂S