Order Entry
Czech Republic
ContactUsLinkComponent
3-[4-Methyl-5-(1H-pyrazol-3-yl)-1,3-thiazol-2-yl]pyridine
3-[4-Methyl-5-(1H-pyrazol-3-yl)-1,3-thiazol-2-yl]pyridine
Catalog # APOSOR183435-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[4-Methyl-5-(1H-pyrazol-3-yl)-1,3-thiazol-2-yl]pyridine
Catalog # APOSOR183435-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :242.3 g/mol
Storage Temperature:Ambient
CAS Number:499796-05-3
Formula:C₁₂H₁₀N₄S
MDL Number:MFCD06808800