Order Entry
Czech Republic
ContactUsLinkComponent
2,7-Dimethyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine
2,7-Dimethyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine
Catalog # APOSOR183433-1G
Supplier:  Apollo Scientific
CAS Number:  
2,7-Dimethyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine
Catalog # APOSOR183433-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :212.25 g/mol
Storage Temperature:Ambient
CAS Number:372198-31-7
Formula:C₁₂H₁₂N₄
MDL Number:MFCD00269378