
N-(2-Hydroxyethyl)methacrylamide 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5238-56-2
:
Formula:C₆H₁₁NO₂
MDL Number:MFCD00080529
:129.157 g/mol
Density:0.95 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together