
4-(4-Bromo-1,3-thiazol-2-yl)morpholine 97%
Catalog # APOSOR16542-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:249.128 g/mol
Melting Pt:74…76 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1017781-60-0
Formula:C₇H₉BrN₂OS
MDL Number:MFCD09607696