Order Entry
Czech Republic
ContactUsLinkComponent
 
 
Naphth-1-ylglyoxal hydrate 95%
  APOSOR1617-100MG
 :  
 
Naphth-1-ylglyoxal hydrate 95%
  APOSOR1617-100MG
 :  

 

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

 

 

 :
 :202.206 g/mol
 :401 °C (760 mm Hg)
 :196 °C
 :GHS07
 :Refrigerator
 :16208-20-1
 :C₁₂H₈O₂·H₂O
 :MFCD08705867