
Tetrahydro-2H-thiopyran-4-carboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:89489-53-2
:
Formula:C₆H₁₀O₂S
MDL Number:MFCD09879921
:146.207 g/mol
Melting Pt:111…112.5 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together