
Methyl 2-chloro-1,3-oxazole-5-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:934236-41-6
:
Formula:C₅H₄ClNO₃
MDL Number:MFCD09265527
:161.543 g/mol
:GHS07
Storage Temperature:Freezer
Frequently Bought Together