
4-(Chloromethyl)-1H-imidazole hydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:38585-61-4
:
Formula:C₄H₅ClN₂·ClH
MDL Number:MFCD01721661
:153.01 g/mol
Melting Pt:139…145 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together