Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl (E)-2-cyano-3-(4-methoxyphenyl)acrylate
Methyl (E)-2-cyano-3-(4-methoxyphenyl)acrylate
Methyl (E)-2-cyano-3-(4-methoxyphenyl)acrylate
Methyl (E)-2-cyano-3-(4-methoxyphenyl)acrylate
APOSOR15502-1G
:
Apollo Scientific
:
Methyl (E)-2-cyano-3-(4-methoxyphenyl)acrylate
APOSOR15502-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
217.221 g/mol
:
GHS07
:
Ambient
:
14479-58-4
:
C₁₂H₁₁NO₃
:
MFCD00630908