
3-Chloro-6-(2-methoxyphenyl)pyridazine
Catalog # APOSOR14962-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:220.655 g/mol
Melting Pt:122 °C
:GHS07
Storage Temperature:Ambient
CAS Number:949468-94-4
Formula:C₁₁H₉ClN₂O
MDL Number:MFCD06801341