
4-Amino-6,7-dimethoxy-2-(piperazin-1-yl)quinazoline
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:60547-97-9
:
Formula:C₁₄H₁₉N₅O₂
MDL Number:MFCD00563872
:289.333 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together