
1,4'-Bipiperidine-1'-carboximidamide hydroiodide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:849776-34-7
:
Formula:C₁₁H₂₂N₄·HI
MDL Number:MFCD07368562
:338.232 g/mol
Melting Pt:180…182 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together