Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 5-(fur-2-yl)-1H-pyrazole-3-carboxylate
Irritant
Ethyl 5-(fur-2-yl)-1H-pyrazole-3-carboxylate
Catalog # APOSOR1451-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl 5-(fur-2-yl)-1H-pyrazole-3-carboxylate
Catalog # APOSOR1451-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :206.198 g/mol
Melting Pt:137…138 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:34020-22-9
Formula:C₁₀H₁₀N₂O₃
MDL Number:MFCD06797478