
[2-(1H-Pyrrol-1-yl)phenyl]methanol 97%
Catalog # APOSOR1431-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:173.211 g/mol
Boiling Pt:104 °C (0.5 mm Hg)
Melting Pt:42…44 °C
:GHS07
Storage Temperature:Ambient
CAS Number:61034-86-4
Formula:C₁₁H₁₁NO
MDL Number:MFCD04115123