Order Entry
Czech Republic
ContactUsLinkComponent
[3-(2-Methyl-1,3-thiazol-4-yl)phenyl]methanol 97%
Irritant
[3-(2-Methyl-1,3-thiazol-4-yl)phenyl]methanol 97%
Catalog # APOSOR1426-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
[3-(2-Methyl-1,3-thiazol-4-yl)phenyl]methanol 97%
Catalog # APOSOR1426-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :205.276 g/mol
Melting Pt:77.5…79.5 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:850375-06-3
Formula:C₁₁H₁₁NOS
MDL Number:MFCD02682066