Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(3-Iodobenzyl)piperazine, N1-BOC protected
4-(3-Iodobenzyl)piperazine, N1-BOC protected 97%
4-(3-Iodobenzyl)piperazine, N1-BOC protected 97%
APOSOR1425-1G
:
Apollo Scientific
:
4-(3-Iodobenzyl)piperazine, N1-BOC protected 97%
APOSOR1425-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
402.27 g/mol
:
72…74 °C
:
Ambient
:
3259
:
8,III
:
GHS05
:
GHS07
:
850375-09-6
:
C₁₆H₂₃IN₂O₂
:
MFCD06658978