cis-3-Azabicyclo[3.1.0]hexane-2-carboxylic acid
Catalog # APOSOR14181-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:127.141 g/mol
Melting Pt:252…256 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1279569-17-3
Formula:C₆H₉NO₂
MDL Number:MFCD03093875