Ethyl 2-cyano-3-(4-methoxyphenyl)acrylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2017-87-0
:2286-29-5
Formula:C₁₃H₁₃NO₃
MDL Number:MFCD00026849
:231.247 g/mol
Melting Pt:85…87 °C
Storage Temperature:Ambient
Frequently Bought Together