
2-(Hydroxymethyl)indoline
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:138969-57-0
:
Formula:C₉H₁₁NO
MDL Number:MFCD06200868
:149.19 g/mol
Melting Pt:49…58 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together