
5,7-Dimethylpyrazolo[1,5-a]pyrimidine-2-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:300691-07-0
:
Formula:C₉H₉N₃O₂
MDL Number:MFCD00461394
:191.187 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together