Order Entry
Czech Republic
ContactUsLinkComponent
4-(2-Methoxyphenoxy)benzoic acid
Irritant
4-(2-Methoxyphenoxy)benzoic acid
Catalog # APOSOR12951-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-(2-Methoxyphenoxy)benzoic acid
Catalog # APOSOR12951-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :244.243 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:103203-54-9
Formula:C₁₄H₁₂O₄
MDL Number:MFCD01570262