
[3-(4-Methoxyphenoxy)phenyl]methylamine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:154108-33-5
:
Formula:C₁₄H₁₅NO₂·ClH
MDL Number:MFCD03840098
:265.735 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together