
4'-Methyl-[1,1'-biphenyl]-2-sulphonyl chloride
Catalog # APOSOR12791-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:266.743 g/mol
:GHS05
Storage Temperature:Ambient
CAS Number:173253-46-8
Formula:C₁₃H₁₁ClO₂S
MDL Number:MFCD03840035